Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170433.1 | NP_001163904.1 | 22 | 2466 | ||
NM_026221.2 | NP_080497.1 | 21 | 2433 | |||
GenBank mRNA | AK014559.1 | - | 5 | 599 | ||
AK044496.1 | - | 14 | 3221 | |||
AK146575.1 | - | 21 | 2417 | |||
AK154482.1 | - | 21 | 2378 | |||
AK173133.1 | - | 22 | 2371 | |||
BC035209.1 | - | 21 | 2279 | |||
BC058176.1 | - | 9 | 969 | |||
BC060078.1 | - | 12 | 2706 | |||
BC071257.1 | - | 2 | 141 | |||
BC072603.1 | - | 21 | 2413 | 6830850 |