Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042499.1 | NP_001035964.1 | 2, 3 | 164 | ||
GenBank mRNA | AK011280.1 | - | 2, 3 | 159 | ||
AK016099.1 | - | 2, 3 | 173 | |||
AK028195.1 | - | 2, 3 | 158 | |||
AK028269.1 | - | 2, 3 | 150 | |||
AK156982.1 | - | 2, 3 | 155 | |||
BC050194.1 | - | 2, 3 | 145 | 30022531 |