Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026443.4 | NP_080719.2 | 3 | 394 | ||
GenBank mRNA | AK006148.1 | - | 3 | 315 | ||
AK010971.1 | - | 3 | 351 | |||
AK012058.1 | - | 3 | 361 | |||
AK078381.1 | - | 3 | 395 | |||
AK167990.1 | - | 3 | 261 | |||
BC019205.1 | - | 3 | 404 | |||
BC058737.1 | - | 3 | 344 | 5400435 |