Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026483.2 | NP_080759.2 | 3 | 871 | ||
GenBank mRNA | AK017490.1 | - | 2 | 1295 | ||
BC018177.1 | - | 3 | 865 | |||
BC038033.1 | - | 3 | 866 | |||
BC049270.1 | - | 3 | 847 | 3988645 | ||
CK390971.1 | - | 2 | 270 |