Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026487.3 | NP_080763.2 | 2 | 139 | ||
GenBank mRNA | AK014967.1 | - | 2 | 156 | ||
AK030719.1 | - | 2 | 159 | |||
AK033639.1 | - | 2 | 163 | |||
AK044925.1 | - | 2 | 139 | |||
AK075685.1 | - | 2 | 183 | |||
AK150469.1 | - | 2 | 159 | |||
AK152059.1 | - | 2 | 159 | |||
AK165953.1 | - | 2 | 174 | |||
BC029085.1 | - | 2 | 139 | 3593451 | ||
BC043051.1 | - | 2 | 124 | 5708377 |