Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD46 SIRNA Assays ›
          Gene Symbol:
          CD46
          Gene Name:
          CD46 molecule
          Gene Aliases:
          AHUS2, MCP, MIC10, TLX, TRA2.10
          Chromosome Location:
          Chr.1: 207752038 - 207795516 on Build GRCh38
          UniGene ID:
          Hs.510402
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s8604

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD46
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002389.4 NP_002380.3 5 702
          NM_153826.3 NP_722548.1 5 702
          NM_172350.2 NP_758860.1 5 702
          NM_172351.2 NP_758861.1 5 702
          NM_172352.2 NP_758862.1 5 702
          NM_172353.2 NP_758863.1 5 702
          NM_172359.2 NP_758869.1 5 702
          NM_172361.2 NP_758871.1 5 702
          XM_011509563.1 XP_011507865.1 5 702
          XM_011509564.1 XP_011507866.1 5 702
          XM_017001308.1 XP_016856797.1 5 702
          XM_017001309.1 XP_016856798.1 5 702
          XM_017001310.1 XP_016856799.1 5 702
          GenBank mRNA A18585.1 - 5 560
          AK222822.1 - 5 628
          AK291227.1 - 5 656
          BC030594.2 - 5 642 4823040
          BX537451.1 - 4 2198
          BX640613.1 - 5 684
          BX649050.1 - 5 673
          D84105.1 - 5 548
          EF076055.1 - 5 560
          EF076056.1 - 5 560
          EF076057.1 - 5 560
          EF076058.1 - 5 560
          M58050.1 - 5 561
          S51940.1 - 5 614
          X59405.1 - 5 672
          X59406.1 - 5 631
          X59407.1 - 5 631
          X59408.1 - 5 672
          X59409.1 - 5 631
          X59410.1 - 5 631
          Y00651.1 - 5 570

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell mediated immunity
          proteolysis
          complement activation, classical pathway
          single fertilization
          regulation of Notch signaling pathway
          positive regulation of gene expression
          negative regulation of gene expression
          regulation of complement activation
          interleukin-10 production
          positive regulation of interleukin-10 production
          sequestering of extracellular ligand from receptor
          positive regulation of T cell proliferation
          negative regulation of catalytic activity
          positive regulation of memory T cell differentiation
          innate immune response
          positive regulation of regulatory T cell differentiation
          negative regulation of complement activation
          viral entry into host cell
          positive regulation of transforming growth factor beta production
          virus receptor activity
          complement binding
          endopeptidase activity
          enzyme inhibitor activity
          receptor activity
          protein binding
          cadherin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0