Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aph1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026674.3 | NP_080950.1 | 3 | 424 | ||
GenBank mRNA | AB197123.1 | - | 3 | 244 | ||
AK002310.1 | - | 3 | 390 | |||
AK155147.1 | - | 3 | 438 | |||
BC050923.1 | - | 3 | 424 | 6410104 | ||
BC063254.1 | - | 3 | 424 | 6410104 |