Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MEIS2 SIRNA Assays ›
Gene Symbol:
MEIS2
Gene Name:
Meis homeobox 2
Gene Aliases:
HsT18361, MRG1
Chromosome Location:
Chr.15: 36891021 - 37101299 on Build GRCh38
UniGene ID:
Hs.510989
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8665
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MEIS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220482.1
NP_001207411.1
7
1171
NM_002399.3
NP_002390.1
6
1026
NM_170674.4
NP_733774.1
6
1304
NM_170675.4
NP_733775.1
6
1304
NM_170676.4
NP_733776.1
6
1304
NM_170677.4
NP_733777.1
6
1304
NM_172315.2
NP_758526.1
5
780
NM_172316.2
NP_758527.1
6
1091
NR_051953.1
6
1304
XM_006720523.1
XP_006720586.1
6
694
XM_006720524.1
XP_006720587.1
6
694
XM_006720525.1
XP_006720588.1
6
694
XM_006720526.2
XP_006720589.1
6
1690
XM_006720527.2
XP_006720590.1
5
1571
XM_006720529.2
XP_006720592.1
5
1571
XM_011521591.2
XP_011519893.1
4
756
XM_017022205.1
XP_016877694.1
6
1690
GenBank mRNA
AF178948.1
-
6
885
AF179896.1
-
6
885
AF179897.1
-
6
885
AF179898.1
-
6
885
AF179899.1
-
6
885
AK055936.1
-
5
504
AK056038.1
-
5
721
AK056216.1
-
6
1703
AK056620.1
-
6
1079
AK226072.1
-
6
1307
AK294411.1
-
4
458
AK300247.1
-
6
1059
BC001516.2
-
6
1030
2964406
BC001844.2
-
6
1030
2964406
BC007202.1
-
6
1030
2964406
BC050431.1
-
7
1051
6066728
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
eye development
transcription from RNA polymerase II promoter
visual learning
response to mechanical stimulus
pancreas development
negative regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
response to growth factor
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
transcription corepressor activity
protein binding
transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0