siRNA Details
Gene Symbol: Micu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028643.3 | NP_082919.1 | 9 | 879 | ||
GenBank mRNA | AK014778.1 | - | 9 | 885 | ||
AK031692.1 | - | 9 | 866 | |||
AK049658.1 | - | 9 | 881 | |||
AK167296.1 | - | 9 | 817 | |||
BC031172.1 | - | 9 | 881 | 4986725 | ||
BC031373.1 | - | 9 | 718 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
