Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAP3K5 SIRNA Assays ›
          Gene Symbol:
          MAP3K5
          Gene Name:
          mitogen-activated protein kinase kinase kinase 5
          Gene Aliases:
          ASK1, MAPKKK5, MEKK5
          Chromosome Location:
          Chr.6: 136557046 - 136793098 on Build GRCh38
          UniGene ID:
          Hs.186486
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s8677

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAP3K5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005923.3 NP_005914.1 1 798
          XM_011535839.2 XP_011534141.2 2 1122
          XM_017010870.1 XP_016866359.1 2 1121
          XM_017010871.1 XP_016866360.1 2 1121
          XM_017010872.1 XP_016866361.1 2 1120
          XM_017010873.1 XP_016866362.1 2 1119
          XM_017010874.1 XP_016866363.1 2 1121
          XM_017010875.1 XP_016866364.1 2 1120
          XM_017010876.1 XP_016866365.1 1 269
          XM_017010877.1 XP_016866366.1 1 506
          GenBank mRNA AB209093.1 - 1 902
          AK310942.1 - 2 577
          BC054503.2 - 1 745 6143181
          BC088829.1 - 1 799 6007002
          D84476.2 - 1 704
          U67156.1 - 1 797

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          response to ischemia
          protein phosphorylation
          JNK cascade
          activation of JUN kinase activity
          intrinsic apoptotic signaling pathway in response to oxidative stress
          viral process
          response to endoplasmic reticulum stress
          p38MAPK cascade
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of JUN kinase activity
          innate immune response
          positive regulation of myoblast differentiation
          positive regulation of JNK cascade
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to hydrogen peroxide
          apoptotic signaling pathway
          programmed necrotic cell death
          positive regulation of neuron death
          cellular response to reactive nitrogen species
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          protein binding
          ATP binding
          protein kinase binding
          protein phosphatase binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0