Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps18c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026826.1 | NP_081102.1 | 4, 5 | 304 | ||
GenBank mRNA | AK004139.1 | - | 4, 5 | 317 | ||
AK088290.1 | - | 4, 5 | 317 | |||
AK145935.1 | - | 4, 5 | 321 | |||
BC028442.1 | - | 4, 5 | 304 | 1196057 |