Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam57b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146347.1 | NP_001139819.1 | 4 | 977 | ||
NM_026884.1 | NP_081160.1 | 8 | 1090 | |||
NM_029978.1 | NP_084254.1 | 5 | 1529 | |||
GenBank mRNA | AK131642.1 | - | 5 | 1528 | ||
BC055444.1 | - | 4 | 978 | 4502433 | ||
BC093505.1 | - | 1115 | 4503230 |