Search
Search

MGAT5| Gene Symbol: MGAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002410.4 | NP_002401.1 | 8 | 1216 | ||
| XM_005263668.4 | XP_005263725.1 | 9 | 1581 | |||
| XM_005263669.4 | XP_005263726.1 | 9 | 1578 | |||
| XM_005263670.3 | XP_005263727.1 | 9 | 1234 | |||
| XM_006712534.3 | XP_006712597.1 | 10 | 1430 | |||
| XM_011511199.2 | XP_011509501.1 | 10 | 1865 | |||
| XM_011511200.1 | XP_011509502.1 | 9 | 1230 | |||
| XM_011511201.2 | XP_011509503.1 | 9 | 1334 | |||
| XM_011511202.1 | XP_011509504.1 | 9 | 1231 | |||
| XM_011511204.2 | XP_011509506.1 | 10 | 1433 | |||
| XM_017004147.1 | XP_016859636.1 | 10 | 1705 | |||
| XM_017004148.1 | XP_016859637.1 | 9 | 1542 | |||
| XM_017004149.1 | XP_016859638.1 | 9 | 1583 | |||
| GenBank mRNA | AF113921.1 | - | 8 | 1104 | ||
| D17716.1 | - | 8 | 1216 | |||