Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178616.3 | NP_848731.2 | 3 | 327 | ||
GenBank mRNA | AK007547.1 | - | 3 | 296 | ||
AK012951.1 | - | 3 | 336 | |||
AK051738.1 | - | 3 | 317 | |||
AK076002.1 | - | 3 | 315 | |||
AK089942.1 | - | 3 | 292 | |||
AK136049.1 | - | 3 | 290 | |||
AK140773.1 | - | 311 | ||||
AK167559.1 | - | 3 | 298 | |||
AK168289.1 | - | 3 | 319 | |||
BC037590.1 | - | 3 | 276 | |||
BC055457.1 | - | 3 | 259 | |||
BC090664.1 | - | 3 | 279 | 30677672 | ||
BC090980.1 | - | 3 | 294 | 30283128 | ||
BC100595.1 | - | 3 | 289 | 6767671 | ||
BC119136.1 | - | 3 | 298 | 8733885 | ||
BC119138.1 | - | 3 | 298 | 8733887 | ||
BY092952.1 | - | 3 | 328 |