siRNA Details
Gene Symbol: Ccdc54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027046.3 | NP_081322.1 | 1 | 1197 | ||
GenBank mRNA | AK005741.1 | - | 1 | 1133 | ||
BC049741.1 | - | 1 | 1197 | 6771599 |

Gene Symbol: Ccdc54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027046.3 | NP_081322.1 | 1 | 1197 | ||
GenBank mRNA | AK005741.1 | - | 1 | 1133 | ||
BC049741.1 | - | 1 | 1197 | 6771599 |