Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2300005B03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081961.1 | NP_001075430.1 | 3 | 250 | ||
GenBank mRNA | AB272582.1 | - | 3 | 238 | ||
AK009031.1 | - | 2 | 173 | |||
BC115610.1 | - | 3 | 250 | 40102714 | ||
BC115611.1 | - | 3 | 250 | 40102716 |