Search
Search

Klhdc2| Gene Symbol: Klhdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027117.3 | NP_081393.2 | 5 | 807 | ||
| GenBank mRNA | AK009481.1 | - | 5 | 722 | ||
| AK051418.1 | - | 5 | 734 | |||
| AK164223.1 | - | 3 | 183 | |||
| AY513272.1 | - | 5 | 758 | |||
| BC005581.1 | - | 5 | 731 | 3968615 | ||