Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027410.1 | NP_081686.1 | 10 | 1355 | ||
GenBank mRNA | AB093295.1 | - | 9 | 1524 | ||
AK032858.1 | - | 10 | 1329 | |||
AK034020.1 | - | 9 | 1933 | |||
AK169888.1 | - | 10 | 1302 | |||
AK170108.1 | - | 10 | 1302 | |||
AK177813.1 | - | 3 | 246 | |||
AK186825.1 | - | 2 | 115 | |||
AK188059.1 | - | 2 | 115 | |||
AK193764.1 | - | 2 | 115 | |||
AK201195.1 | - | 3 | 245 | |||
AK203130.1 | - | 2 | 115 | |||
AK218769.1 | - | 2 | 115 | |||
BC046968.1 | - | 10 | 1355 | 6306206 | ||
BC052185.1 | - | 4 | 1932 |