Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asnsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290984.1 | NP_001277913.1 | 5 | 567 | ||
NM_133728.3 | NP_598489.2 | 5 | 1998 | |||
GenBank mRNA | AK031295.1 | - | 5 | 1956 | ||
AK034746.1 | - | 1917 | ||||
AK049194.1 | - | 5 | 1945 | |||
AK050045.1 | - | 5 | 1938 | |||
AK076793.1 | - | 5 | 1900 | |||
AK155030.1 | - | 5 | 1937 | |||
AK168103.1 | - | 5 | 1938 | |||
AK168534.1 | - | 5 | 568 | |||
AK170510.1 | - | 5 | 1914 | |||
AK176998.1 | - | 1 | 41 | |||
AK205803.1 | - | 1 | 22 | |||
AK218145.1 | - | 1 | 23 | |||
BC013617.1 | - | 5 | 1818 | 3979452 |