Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027475.3 | NP_081751.1 | 10, 11 | 1579 | ||
GenBank mRNA | AK017681.1 | - | 8, 9 | 1003 | ||
AK028664.1 | - | 10, 11 | 1543 | |||
AK043261.1 | - | 10, 11 | 1428 | |||
AK202761.1 | - | 1, 2 | 58 | |||
BC013470.1 | - | 10, 11 | 1528 | 4220504 |