Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195496.1 | NP_001182425.1 | 2 | 1392 | ||
NM_001195497.1 | NP_001182426.1 | 2 | 1392 | |||
NM_001195498.1 | NP_001182427.1 | 2 | 1392 | |||
NM_001195499.1 | NP_001182428.1 | 2 | 1392 | |||
NM_001195500.1 | NP_001182429.1 | 2 | 1392 | |||
NM_027539.5 | NP_081815.3 | 2 | 1392 | |||
GenBank mRNA | AB198721.1 | - | 2 | 749 | ||
AK049126.1 | - | 2 | 882 | |||
AK049179.1 | - | 2 | 844 | |||
AK082633.1 | - | 2 | 888 | |||
AY968049.1 | - | 2 | 680 | |||
AY968050.1 | - | 2 | 680 | |||
AY968051.1 | - | 2 | 680 | |||
BC056921.1 | - | 2 | 1360 | 6822681 | ||
BC060192.1 | - | 2 | 696 |