Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168223.1 | NP_001161695.1 | 6 | 612 | ||
NM_001168224.1 | NP_001161696.1 | 6 | 724 | |||
NM_027562.4 | NP_081838.1 | 7 | 786 | |||
GenBank mRNA | AB091386.1 | - | 4 | 418 | ||
AK014570.1 | - | 7 | 787 | |||
AK137753.1 | - | 6 | 722 | |||
AY137341.1 | - | 6 | 724 | |||
AY137342.1 | - | 6 | 612 | |||
BC117543.1 | - | 7 | 776 | 40105656 | ||
JF431014.1 | - | 5 | 564 | |||
JF792238.1 | - | 6 | 623 | |||
JF792239.1 | - | 6 | 674 |