Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CD200 SIRNA Assays ›
Gene Symbol:
CD200
Gene Name:
CD200 molecule
Gene Aliases:
MOX1, MOX2, MRC, OX-2
Chromosome Location:
Chr.3: 112332224 - 112362812 on Build GRCh38
UniGene ID:
Hs.79015
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8925
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CD200
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001004196.3
NP_001004196.2
4
614
NM_001318826.1
NP_001305755.1
3
539
NM_001318828.1
NP_001305757.1
2
457
NM_001318830.1
NP_001305759.1
2
457
NM_005944.6
NP_005935.4
3
539
XM_005247481.4
XP_005247538.1
4
788
XM_005247482.2
XP_005247539.1
3
418
XM_005247485.2
XP_005247542.1
3
729
XM_005247486.2
XP_005247543.1
2
226
XM_005247488.2
XP_005247545.1
2
336
XM_017006452.1
XP_016861941.1
4
1115
XM_017006453.1
XP_016861942.1
2
381
XM_017006454.1
XP_016861943.1
4
1104
XM_017006455.1
XP_016861944.1
3
739
XM_017006456.1
XP_016861945.1
4
465
XM_017006457.1
XP_016861946.1
3
418
XM_017006458.1
XP_016861947.1
4
1110
XM_017006459.1
XP_016861948.1
3
729
GenBank mRNA
AF063591.1
-
3
330
AK074972.1
-
3
304
AK129797.1
-
2
226
AK293399.1
-
2
226
AK297194.1
-
4
405
AK312563.1
-
3
420
AL832529.1
-
3
324
AY603771.1
-
2
273
BC022522.1
-
3
332
4794050
BC031103.1
-
3
308
5301702
BI547787.1
-
4
410
DA544134.1
-
3
417
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell recognition
negative regulation of macrophage activation
regulation of immune response
receptor activity
receptor binding
protein binding
protein homodimerization activity
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0