Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MST1 SIRNA Assays ›
          Gene Symbol:
          MST1 LOC102724562 MST1P2
          Gene Name:
          macrophage stimulating 1
          macrophage stimulating 1 pseudogene
          macrophage stimulating 1 pseudogene 2
          Gene Aliases:
          D3F15S2, DNF15S2, HGFL, MSP, NF15S2
          -
          MSPL-1, MSPL-2, MSPL-3, MSPL1, MSPL2, MSPL3, MSTP1, MSTP2, MSTP3
          Chromosome Location:
          Chr.3: 49683947 - 49689528 on Build GRCh38
          Chromosome Location:
          Chr.1|NT_187368: 4205 - 7201 on Build GRCh38
          Chromosome Location:
          Chr.1: 16645574 - 16650420 on Build GRCh38
          UniGene ID:
          Hs.349110
          Hs.349110
          Hs.349110
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s8995

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MST1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_020998.3 NP_066278.3 14 1659
          XM_006713166.1 XP_006713229.1 13 2289
          XM_011533732.1 XP_011532034.1 14 2460
          XM_011533737.2 XP_011532039.1 14 2420
          XM_011533738.2 XP_011532040.1 14 2464
          XM_017006460.1 XP_016861949.1 14 2460
          XM_017006461.1 XP_016861950.1 14 2416
          XM_017006462.1 XP_016861951.1 14 2460
          XM_017006463.1 XP_016861952.1 14 2460
          XM_017006464.1 XP_016861953.1 14 2460
          XM_017006465.1 XP_016861954.1 14 2460
          XM_017006466.1 XP_016861955.1 14 2460
          XM_017006467.1 XP_016861956.1 14 2456
          GenBank mRNA AK222893.1 - 14 1550
          BC044862.1 - 9 3524
          BC048330.1 - 14 1558 5190966
          L11924.1 - 14 1555
          M74178.1 - 14 1545
          Gene Symbol: LOC102724562
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_135824.1 7 1036
          GenBank mRNA AK307907.1 - 5 1219
          AK311553.1 - 2 1722
          Gene Symbol: MST1P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027504.1 10 1860
          GenBank mRNA AK125231.1 - 3 1507

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          biological_process
          negative regulation of gluconeogenesis
          hepatocyte growth factor receptor signaling pathway
          regulation of cAMP-dependent protein kinase activity
          molecular_function
          serine-type endopeptidase activity
          protein binding
          receptor tyrosine kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0