Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027895.1 | NP_082171.1 | 3 | 436 | ||
GenBank mRNA | AK004783.1 | - | 3 | 420 | ||
AK032030.1 | - | 1 | 57 | |||
AK154642.1 | - | 3 | 349 | |||
BC037093.1 | - | 3 | 457 | |||
BC151153.1 | - | 3 | 362 | 9088047 |