Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028095.1 | NP_082371.1 | 2 | 161 | ||
GenBank mRNA | AK008476.1 | - | 2 | 146 | ||
BC038323.1 | - | 2 | 116 | |||
BC051506.1 | - | 2 | 136 | |||
BC096622.1 | - | 2 | 145 | 30445095 | ||
BC116375.2 | - | 2 | 159 | 40101137 | ||
BQ830246.1 | - | 2 | 186 |