Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303253.1 | NP_001290182.1 | 8 | 1190 | ||
NM_005891.2 | NP_005882.2 | 8 | 1075 | |||
GenBank mRNA | AB208993.1 | - | 8 | 965 | ||
AF356877.1 | - | 8 | 998 | |||
AK055001.1 | - | 3 | 1174 | |||
AK225089.1 | - | 8 | 976 | |||
AK225244.1 | - | 8 | 988 | |||
AK291080.1 | - | 8 | 976 | |||
AK294273.1 | - | 8 | 1190 | |||
BC000408.2 | - | 8 | 955 | 2823036 | ||
S70154.1 | - | 8 | 981 |