Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028195.3 | NP_082471.2 | 10 | 1005 | ||
GenBank mRNA | AF079972.1 | - | 10 | 936 | ||
AK030956.1 | - | 10 | 1890 | |||
AK152306.1 | - | 10 | 911 | |||
AK152909.1 | - | 10 | 911 | |||
AK157026.1 | - | 10 | 911 | |||
AK220198.1 | - | 10 | 934 | |||
BC018505.1 | - | 9 | 829 | |||
BC039640.1 | - | 3 | 424 | |||
BC139054.1 | - | 10 | 978 | 8862076 | ||
BC139056.1 | - | 10 | 978 | 8862078 | ||
BC145528.1 | - | 10 | 1005 |