Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calhm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133746.5 | NP_598507.3 | 2 | 719 | ||
GenBank mRNA | AK012923.1 | - | 2 | 646 | ||
AK040739.1 | - | 2 | 720 | |||
AK089278.1 | - | 2 | 720 | |||
AK170068.1 | - | 2 | 690 | |||
BC019197.1 | - | 2 | 641 | 5037829 |