Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028379.5 | NP_082655.1 | 6 | 674 | ||
GenBank mRNA | AK007572.1 | - | 4 | 660 | ||
AK013609.1 | - | 6 | 661 | |||
AK156167.1 | - | 6 | 657 | |||
BC019523.1 | - | 6 | 656 | 4163453 |