Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700001O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198000.3 | NP_932117.2 | 4, 5 | 894 | ||
GenBank mRNA | AK005618.1 | - | 4, 5 | 405 | ||
AK007200.1 | - | 383 | ||||
AK076928.1 | - | 4, 5 | 395 | |||
BC023044.1 | - | 4, 5 | 864 | 5355351 |