Search
Search

Capn9| Gene Symbol: Capn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023709.4 | NP_076198.2 | 11 | 1386 | ||
| GenBank mRNA | AK008629.1 | - | 11 | 1387 | ||
| AK144514.1 | - | 11 | 1389 | |||
| BC058748.1 | - | 11 | 1364 | 6394969 | ||
| U89513.1 | - | 11 | 1347 | |||