Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028627.2 | NP_082903.2 | 18 | 3090 | ||
GenBank mRNA | AK003542.1 | - | 2 | 65 | ||
AK158685.1 | - | 8 | 896 | |||
AK158851.1 | - | 8 | 1022 | |||
AK220438.1 | - | 13 | 3207 | |||
BC043135.1 | - | 14 | 1919 | |||
BC057353.1 | - | 18 | 3091 | |||
BC058352.1 | - | 4 | 465 | |||
BC138652.1 | - | 16 | 2949 | 8861672 | ||
BC145352.1 | - | 16 | 2952 | 9053908 |