Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103156.2 | NP_001096626.1 | 4 | 994 | ||
NM_001103157.2 | NP_001096627.1 | 4 | 1349 | |||
NM_001285469.1 | NP_001272398.1 | 4 | 903 | |||
NM_001285470.1 | NP_001272399.1 | 3 | 868 | |||
NM_001285471.1 | NP_001272400.1 | 3 | 868 | |||
NM_028734.5 | NP_083010.2 | 4 | 1051 | |||
GenBank mRNA | AK052981.1 | - | 4 | 1053 | ||
AK086787.1 | - | 4 | 1022 | |||
AK089927.1 | - | 3 | 842 | |||
AK145468.1 | - | 4 | 940 | |||
AK162343.1 | - | 3 | 830 | |||
AK165644.1 | - | 4 | 1314 | |||
AK166162.1 | - | 4 | 1018 | |||
AK166607.1 | - | 4 | 940 | |||
BC043459.1 | - | 4 | 904 | |||
BC150881.1 | - | 4 | 910 | 9087792 | ||
BC150889.1 | - | 4 | 910 | 9087800 | ||
BC150997.1 | - | 961 | 9087910 | |||
BC151008.1 | - | 961 | 9087921 |