Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163590.1 | NP_001157062.1 | 2 | 1009 | ||
NM_001163591.1 | NP_001157063.1 | 2 | 894 | |||
NM_029075.1 | NP_083351.1 | 3 | 1144 | |||
GenBank mRNA | AK017897.1 | - | 3 | 967 | ||
AK153408.1 | - | 3 | 1119 | |||
AK171040.1 | - | 2 | 927 | |||
AK171663.1 | - | 935 | ||||
BC117539.1 | - | 2 | 847 | 40105862 | ||
BC118516.1 | - | 2 | 848 | 40105861 |