Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029238.2 | NP_083514.1 | 4 | 876 | ||
GenBank mRNA | AB041538.1 | - | 5 | 804 | ||
AK034308.1 | - | 4 | 878 | |||
AK046093.1 | - | 4 | 1086 | |||
AK047949.1 | - | 4 | 873 | |||
BC138981.1 | - | 4 | 775 | 8862003 | ||
BC145720.1 | - | 4 | 775 | 40131023 |