Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SIRT4 SIRNA Assays ›
Gene Symbol:
Sirt4
Gene Name:
sirtuin 4
Gene Aliases:
4930596O17Rik
Chromosome Location:
Chr.5: 115478006 - 115490050 on Build GRCm38
UniGene ID:
Mm.332616
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s93444
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Sirt4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_133760.1
NP_598521.1
1
102
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein ADP-ribosylation
glutamine metabolic process
cellular response to DNA damage stimulus
negative regulation of cardiac muscle cell apoptotic process
peptidyl-lysine deacetylation
negative regulation of fatty acid oxidation
negative regulation of insulin secretion
positive regulation of lipid biosynthetic process
tricarboxylic acid metabolic process
negative regulation of protein processing involved in protein targeting to mitochondrion
imidazoleglycerol-phosphate synthase activity
NAD+ ADP-ribosyltransferase activity
histone deacetylase activity
protein binding
protein-N-terminal asparagine amidohydrolase activity
UDP-3-O-[3-hydroxymyristoyl] N-acetylglucosamine deacetylase activity
transferase activity
hydrolase activity
NAD(P)-cysteine ADP-ribosyltransferase activity
NAD(P)-asparagine ADP-ribosyltransferase activity
NAD(P)-serine ADP-ribosyltransferase activity
iprodione amidohydrolase activity
(3,5-dichlorophenylurea)acetate amidohydrolase activity
4'-(2-hydroxyisopropyl)phenylurea amidohydrolase activity
didemethylisoproturon amidohydrolase activity
N-isopropylacetanilide amidohydrolase activity
N-cyclohexylformamide amidohydrolase activity
isonicotinic acid hydrazide hydrolase activity
cis-aconitamide amidase activity
gamma-N-formylaminovinylacetate hydrolase activity
NAD-dependent protein deacetylase activity
N2-acetyl-L-lysine deacetylase activity
O-succinylbenzoate synthase activity
indoleacetamide hydrolase activity
7-cyano-7-deazaguanine tRNA-ribosyltransferase activity
N-acetylcitrulline deacetylase activity
purine deoxyribosyltransferase activity
metal ion binding
N-acetylgalactosamine-6-phosphate deacetylase activity
diacetylchitobiose deacetylase activity
chitooligosaccharide deacetylase activity
NAD+ binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0