Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ascc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029291.1 | NP_083567.1 | 5, 6 | 586 | ||
GenBank mRNA | AK048876.1 | - | 5, 6 | 620 | ||
AK051387.1 | - | 5, 6 | 635 | |||
AK148105.1 | - | 5, 6 | 618 | |||
AK212915.1 | - | 1, 2 | 71 | |||
BC013537.1 | - | 5, 6 | 586 | 4240784 |
Gene Symbol: Dcaf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK179635.1 | - | 96 |