Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168331.1 | NP_001161803.1 | 1 | 206 | ||
NM_004547.5 | NP_004538.2 | 1 | 206 | |||
GenBank mRNA | AF044957.1 | - | 1 | 135 | ||
BC000855.1 | - | 1 | 178 | 3459215 | ||
BC070285.1 | - | 1 | 145 | 6714676 | ||
BC146885.1 | - | 1 | 177 | 9056813 | ||
BC146887.1 | - | 1 | 177 | 9056815 | ||
BC146927.1 | - | 1 | 177 | 9056855 | ||
BC146932.1 | - | 1 | 177 | 9056860 | ||
BF212059.1 | - | 202 | ||||
BF978138.1 | - | 206 |