Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RAPGEF2 SIRNA Assays ›
Gene Symbol:
Rapgef2
Gene Name:
Rap guanine nucleotide exchange factor (GEF) 2
Gene Aliases:
5830453M24Rik, CNRasGEF, Pdzgef1, RA-GEF-1, mKIAA0313, nRapGEP
Chromosome Location:
Chr.3: 79062516 - 79287244 on Build GRCm38
UniGene ID:
Mm.31220
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s94079
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rapgef2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099624.2
NP_001093094.2
4
472
GenBank mRNA
AK147368.1
-
4
400
BC067056.1
-
4
472
EF410089.1
-
621
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
neuron migration
signal transduction
G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
small GTPase mediated signal transduction
multicellular organism development
nervous system development
negative regulation of cell proliferation
positive regulation of neuron projection development
cAMP-mediated signaling
ventricular system development
forebrain neuron development
microvillus assembly
cell differentiation
neuron projection development
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of protein binding
Rap protein signal transduction
nerve growth factor signaling pathway
positive regulation of GTPase activity
positive regulation of cAMP-mediated signaling
positive regulation of protein kinase activity
negative regulation of melanin biosynthetic process
regulation of synaptic plasticity
negative regulation of dendrite morphogenesis
establishment of endothelial barrier
positive regulation of ERK1 and ERK2 cascade
cellular response to cAMP
cellular response to cGMP
adenylate cyclase-activating adrenergic receptor signaling pathway
protein localization to plasma membrane
establishment of endothelial intestinal barrier
regulation of cell junction assembly
cellular response to nerve growth factor stimulus
positive regulation of cAMP-dependent protein kinase activity
positive regulation of dendritic cell apoptotic process
positive regulation of vasculogenesis
positive regulation of neuron migration
guanyl-nucleotide exchange factor activity
Ras guanyl-nucleotide exchange factor activity
GTPase activator activity
Rap guanyl-nucleotide exchange factor activity
protein kinase binding
PDZ domain binding
beta-1 adrenergic receptor binding
WW domain binding
phosphatidic acid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System