Search
Search

Fads1| Gene Symbol: Fads1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146094.2 | NP_666206.1 | 11 | 1446 | ||
| GenBank mRNA | AB072976.1 | - | 11 | 1361 | ||
| AF061504.1 | - | 11 | 1347 | |||
| AK033308.1 | - | 9 | 803 | |||
| AK083959.1 | - | 11 | 1308 | |||
| AK154367.1 | - | 11 | 1396 | |||
| BC013226.1 | - | 1 | 50 | |||
| BC022139.1 | - | 11 | 1391 | 5100199 | ||
| BC026831.1 | - | 11 | 1413 | 4239695 | ||
| BC026848.1 | - | 11 | 1386 | 4196030 | ||
| BC063053.1 | - | 11 | 1306 | 5696722 | ||
| KF987079.1 | - | 11 | 1330 | |||