Search
Search

Fads1| Gene Symbol: Fads1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146094.2 | NP_666206.1 | 2 | 466 | ||
| GenBank mRNA | AB072976.1 | - | 2 | 381 | ||
| AF061504.1 | - | 2 | 368 | |||
| AK080706.1 | - | 2 | 370 | |||
| AK083959.1 | - | 2 | 328 | |||
| AK154367.1 | - | 2 | 416 | |||
| BC022139.1 | - | 2 | 411 | 5100199 | ||
| BC026831.1 | - | 2 | 433 | 4239695 | ||
| BC026848.1 | - | 2 | 406 | 4196030 | ||
| BC063053.1 | - | 2 | 326 | 5696722 | ||
| KF987079.1 | - | 2 | 350 | |||