Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gid8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289651.1 | NP_001276580.1 | 5 | 762 | ||
NM_001289652.1 | NP_001276581.1 | 5 | 728 | |||
NM_029607.2 | NP_083883.1 | 4 | 1248 | |||
GenBank mRNA | AK009106.1 | - | 5 | 688 | ||
AK213539.1 | - | 1 | 27 | |||
BC052539.1 | - | 5 | 760 | |||
BC059022.1 | - | 4 | 1248 | 6415235 |