Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap4-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029613.1 | NP_083889.1 | 1 | 845 | ||
GenBank mRNA | AK009665.1 | - | 1 | 845 | ||
BC115508.1 | - | 1 | 820 | 40101065 | ||
BC115509.1 | - | 1 | 820 | 40101067 |