Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GLT8D1 SIRNA Assays ›
          Gene Symbol:
          Glt8d1
          Gene Name:
          glycosyltransferase 8 domain containing 1
          Gene Aliases:
          2410004H05Rik, 5430414N14Rik, AI450005
          Chromosome Location:
          Chr.14: 31001347 - 31012441 on Build GRCm38
          UniGene ID:
          Mm.8766
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s94392

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Glt8d1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001165930.1 NP_001159402.1 9 975
          NM_029626.2 NP_083902.2 9 1504
          GenBank mRNA AK010388.1 - 9 1233
          AK077388.1 - 9 968
          AK217645.1 - 3 274
          BC029215.1 - 7 1176
          BC055709.1 - 8 1391
          BC058731.1 - 8 1391
          BC069873.1 - 9 1054 6314675

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001007683, AY325256, BC079066.

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          alpha-1,6-mannosyltransferase activity
          alpha-1,2-mannosyltransferase activity
          mannosyltransferase activity
          alpha-1,3-mannosyltransferase activity
          alpha-1,3-galactosyltransferase activity
          molecular_function
          UDP-glucose:glycoprotein glucosyltransferase activity
          glycolipid mannosyltransferase activity
          oligosaccharyl transferase activity
          dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
          acetylglucosaminyltransferase activity
          acetylgalactosaminyltransferase activity
          galactosyltransferase activity
          fucosyltransferase activity
          O antigen polymerase activity
          lipopolysaccharide-1,6-galactosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          cellulose synthase activity
          9-phenanthrol UDP-glucuronosyltransferase activity
          1-phenanthrol glycosyltransferase activity
          9-phenanthrol glycosyltransferase activity
          1,2-dihydroxy-phenanthrene glycosyltransferase activity
          phenanthrol glycosyltransferase activity
          beta-1,4-mannosyltransferase activity
          alpha-1,2-galactosyltransferase activity
          dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
          lipopolysaccharide-1,5-galactosyltransferase activity
          dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
          dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
          inositol phosphoceramide synthase activity
          glucosyltransferase activity
          alpha-(1->3)-fucosyltransferase activity
          alpha-(1->6)-fucosyltransferase activity
          indole-3-butyrate beta-glucosyltransferase activity
          salicylic acid glucosyltransferase (ester-forming) activity
          salicylic acid glucosyltransferase (glucoside-forming) activity
          benzoic acid glucosyltransferase activity
          chondroitin hydrolase activity
          dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
          cytokinin 9-beta-glucosyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline