Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029640.2 | NP_083916.1 | 17 | 2100 | ||
NM_180662.2 | NP_850993.2 | 17 | 2845 | |||
GenBank mRNA | AK156739.1 | - | 18 | 2670 | ||
AK173287.1 | - | 7 | 844 | |||
AY630620.1 | - | 17 | 2098 | |||
BC019994.1 | - | 3 | 330 | |||
BC034590.1 | - | 17 | 2098 | 5364921 | ||
BC038938.1 | - | 11 | 1415 | |||
BC070463.1 | - | 17 | 2845 | 30613261 |