Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MDH1B SIRNA Assays ›
Gene Symbol:
Mdh1b
Gene Name:
malate dehydrogenase 1B, NAD (soluble)
Gene Aliases:
1700124B08Rik, AV255588
Chromosome Location:
Chr.1: 63687573 - 63730547 on Build GRCm38
UniGene ID:
Mm.30494
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s94556
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Mdh1b
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_029696.4
NP_083972.3
8, 9
1374
GenBank mRNA
AK007262.1
-
4, 5
499
BC050786.1
-
8, 9
1374
6703279
Back To Top
More Information
Panther Classification:
Molecular Function -
dehydrogenase
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
tricarboxylic acid cycle
malate metabolic process
oxidation-reduction process
C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
catalytic activity
aldo-keto reductase (NADP) activity
isocitrate dehydrogenase activity
mevaldate reductase activity
gluconate dehydrogenase activity
steroid dehydrogenase activity
oxidoreductase activity
malate dehydrogenase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
epoxide dehydrogenase activity
5-exo-hydroxycamphor dehydrogenase activity
2-hydroxytetrahydrofuran dehydrogenase activity
acetoin dehydrogenase activity
phenylcoumaran benzylic ether reductase activity
D-xylose:NADP reductase activity
L-arabinose:NADP reductase activity
D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
steroid dehydrogenase activity, acting on the CH-CH group of donors
(R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
3-hydroxymenthone dehydrogenase activity
very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
dihydrotestosterone 17-beta-dehydrogenase activity
(R)-2-hydroxyisocaproate dehydrogenase activity
L-arabinose 1-dehydrogenase (NADP+) activity
L-xylulose reductase (NAD+) activity
3-ketoglucose-reductase activity
(R)-2-hydroxyglutarate dehydrogenase activity
D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-xct9l:80/100.66.75.150:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0