Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029749.2 | NP_084025.2 | 16 | 3915 | ||
GenBank mRNA | BC137852.1 | - | 15 | 3808 | 8860871 | |
CV884015.1 | - | 1 | 111 |