Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133972.2 | NP_598733.2 | 8 | 1528 | ||
GenBank mRNA | AK049351.1 | - | 8 | 1244 | ||
AK080609.1 | - | 8 | 1233 | |||
AK081550.1 | - | 8 | 1573 | |||
BC005754.1 | - | 5 | 737 | |||
BC030077.1 | - | 5 | 557 | |||
BC043070.1 | - | 8 | 1528 | 6403587 | ||
BC048396.1 | - | 8 | 1490 | |||
BC055779.1 | - | 8 | 1528 | 6403587 |