Search
Search

Slitrk1| Gene Symbol: Slitrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199065.2 | NP_951020.1 | 1 | 2190 | ||
| GenBank mRNA | AK014285.1 | - | 1 | 126 | ||
| AK144290.1 | - | 1 | 2041 | |||
| AK173296.1 | - | 2109 | ||||
| BC150756.1 | - | 1 | 1298 | 9087667 | ||
| BC157902.1 | - | 1 | 1298 | 9007276 | ||